To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Membrane-associated ring Finger (C3HC4) 2, E3 ubiquitin protein ligase
- Clonality:Polyclonal
- Gene ID:51257
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Antigen symbol:MARCH2
- Conjugation:FITC (Fluorescein Isothiocyanate)
- ImmunoChemistry:Yes
- Size:100 µL
- Cross adsorption:No
- Form:liquid
- Antigen synonyms:RNF172|MARCH-II|HSPC240
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at –20 °C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Cat. no.:10489-062
- Supplier No.:BS-9336R-FITC
Specifications
About this item
Ubiquitination is an important mechanism through which three classes of enzymes act in concert to target short-lived or abnormal proteins for destruction. The three classes of enzymes involved in ubiquitination are the ubiquitin-activating enzymes (E1s), the ubiquitin-conjugating enzymes (E2s) and the ubiquitin-protein ligases (E3s). MARCH2 (membrane-associated ring finger (C3HC4) 2), also known as RNF172 or HSPC240, is a 246 amino acid multi-pass membrane protein that localizes to the endoplasmic reticulum and contains one RING-CH-type zinc finger. Expressed in a variety of tissues, MARCH2 functions as an E3 ubiquitin-protein ligase that is thought to mediate the ubiquitination and subsequent degradation of CD71 and B7-2 and may be involved in endosomal protein trafficking.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: MARCH2
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat