To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Spindle Assembly 6 Homolog (C. elegans)
- Clonality:Polyclonal
- Gene ID:163786
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Antigen symbol:SASS6
- Conjugation:Alexa Fluor® 488
- ImmunoChemistry:Yes
- Size:100 µL
- Cross adsorption:No
- Form:liquid
- Antigen synonyms:MCPH14|SAS-6|SAS6
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at –20 °C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Cat. no.:10486-600
- Supplier No.:BS-9301R-A488
Specifications
About this item
SAS-6 (spindle assembly abnormal protein 6 homolog, HsSAS-6) is a 657 amino acid protein encoded by the human gene SAS6. SAS-6 is a component of the centrosome that contains one PISA (present in SAS-6) domain. LK4, SAS-6, CPAP and other centriole related proteins are required at different stages of procentriole formation and were associated with different centriolar structures. SAS-6 associates only transiently with nascent procentrioles, whereas CEP135 and CPAP form a core structure within the proximal lumen of both parental and nascent centrioles. SAS-6 is necessary for procentriole formation in human cell lines and is localized asymmetrically next to the centriole at the onset of procentriole formation. SAS-6 levels oscillate during the cell cycle; it is degraded in mitosis starting at anaphase, and it accumulates again at the end of the following G1 phase. The anaphase-promoting complex targets SAS-6 for degradation by the 26S Proteasome, and a KEN box in the C-terminus of SAS-6 is necessary for its degradation. Increased SAS-6 levels promoted the formation of multiple procentrioles forming next to a single centriole.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: SASS6
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat