To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Zinc Finger E-box Binding Homeobox 2
- Clonality:Polyclonal
- Gene ID:9839
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Antigen symbol:ZEB2
- Conjugation:Cy3®
- ImmunoChemistry:Yes
- Size:100 µL
- Cross adsorption:No
- Form:liquid
- Antigen synonyms:HSPC082|Smad-interacting protein 1|SIP1|ZFX1B|KIAA0569|Zinc finger E-box-binding homeobox 2|SMADIP1|ZFHX1B|SIP-1|HRIHFB2411|ZEB2
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at –20 °C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Cat. no.:10668-700
- Supplier No.:BS-9186R-CY3
Specifications
About this item
SMAD regulates gene expression by interacting with different classes of transcription factors including DNA-binding multi-zinc finger proteins. SIP1, for SMAD interacting protein 1, is a member of the delta-EF1/Zfh1 family of 2-handed zinc finger/homeodomain proteins. SIP1 contains a SMAD-binding domain, a homeodomain and two clusters of zinc fingers on the N- and C-termini. SIP1, also known as SMADIP1, ZFHX1B and ZEB2 (zinc finger E-box-binding protein 2), can be induced by TGF∫ treatment. SIP1 plays a crucial role in normal embryonic development of neural structures and the neural crest. The human SIP1 gene maps to chromosome 2q22. Mutations in the SIP1 gene cause a form of Hirschsprung disease (HSCR). Patients with SIP1 mutations show mental retardation, delayed motor development, epilepsy, microcephaly, distinct facial features and/or congenital heart disease—all symptoms of HSCR.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: ZEB2
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat