To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Tripartite Motif Containing 50
- Clonality:Polyclonal
- Gene ID:135892
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Antigen symbol:TRIM50
- Conjugation:Cy3®
- ImmunoChemistry:Yes
- Size:100 µL
- Cross adsorption:No
- Form:liquid
- Antigen synonyms:TRIM50A|E3 ubiquitin protein ligase TRIM50|tripartite mot protein 50A|Tripartite mot containing protein 50|E3 ubiquitin ligase|tripartite mot-containing 50A|TRI50_HUMAN.
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at –20 °C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Cat. no.:10668-068
- Supplier No.:BS-9152R-CY3
Specifications
About this item
The tripartite motif (TRIM) family of proteins are characterized by a conserved TRIM domain that includes a coiled-coil region, a B-box type zinc finger, one RING finger and three zinc-binding domains. TRIM50 (tripartite motif containing 50), also known as TRIM50A or E3 ubiquitin-protein ligase TRIM50, is a 487 amino acid cytoplasmic protein that functions as an E3 ubiquitin-protein ligase. Containing one RING-type zinc finger, a B30.2/SPRY domain and a single B box-type zinc finger, TRIM50 belongs to the TRIM/RBCC family and undergoes post-translational auto-ubiquitination. TRIM50 exists as two alternatively spliced isoforms, designated TRIM50 alpha and TRIM50 beta, and has the ability to form dimers and trimers. The gene encoding TRIM50 maps to human chromosome 7, which houses over 1,000 genes, comprises nearly 5% of the human genome and has been linked to Osteogenesis imperfecta, Pendred syndrome, Lissencephaly, Citrullinemia and Shwachman-Diamond syndrome.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: TRIM50
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat