To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Zinc Finger Protein 230
- Clonality:Polyclonal
- Gene ID:50862
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Antigen symbol:ZNF230
- Conjugation:Cy5.5®
- ImmunoChemistry:Yes
- Size:100 µL
- Cross adsorption:No
- Form:liquid
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at –20 °C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Cat. no.:10667-936
- Supplier No.:BS-9143R-CY5.5
Specifications
About this item
The RING-type zinc finger motif is present in a number of viral and eukaryotic proteins and is made of a conserved cysteine-rich domain that is able to bind two zinc atoms. Proteins that contain this conserved domain are generally involved in the ubiquitination pathway of protein degradation. RNF141 (ring finger protein 141), also known as ZFP26 or ZNF230, is a 230 amino acid protein that contains one RING-type zinc finger. Expressed as two isoforms (isoform 1 and isoform 2) due to alternative splicing events, RNF141 is thought to function as a transcription factor during spermatogenesis. While isoform 2 is expressed in brain, heart, pancreas, kidney and skeletal muscle, isoform 1 is expressed primarily in testis, suggesting that isoform 1 functions during spermatogenesis. In addition, RNF141 is not expressed in azoospermic (infertile) men, further implicating an important role for RNF141 in testis development and male fertility.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: ZNF230
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat