To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:COP9 signalosome subunit 8
- Clonality:Polyclonal
- Gene ID:10920
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Antigen symbol:COPS8
- Conjugation:Alexa Fluor® 647
- ImmunoChemistry:Yes
- Size:100 µL
- Cross adsorption:No
- Form:liquid
- Antigen synonyms:SGN8|CSN8|COP9
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at –20 °C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Cat. no.:10667-440
- Supplier No.:BS-9119R-A647
Specifications
About this item
The protein encoded by this gene is one of the eight subunits of COP9 signalosome, a highly conserved protein complex that functions as an important regulator in multiple signaling pathways. The structure and function of COP9 signalosome is similar to that of the 19S regulatory particle of 26S proteasome. COP9 signalosome has been shown to interact with SCF-type E3 ubiquitin ligases and act as a positive regulator of E3 ubiquitin ligases. Alternatively spliced transcript variants encoding distinct isoforms have been observed. [provided by RefSeq, Jul 2008].
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: COPS8
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype:
Reactivity: