To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:ART5
- Clonality:Polyclonal
- Gene ID:116969
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Antigen symbol:ARTC5
- Conjugation:Cy5®
- ImmunoChemistry:Yes
- Size:100 µL
- Cross adsorption:No
- Form:liquid
- Antigen synonyms:ARTC5
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at –20 °C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Cat. no.:10667-180
- Supplier No.:BS-9105R-CY5
Specifications
About this item
Mono-ADP-ribosylation is one of the posttranslational protein modifications regulating cellular metabolism (e.g. nitrogen fixation) in prokaryotes. Mono-ADP-ribosylation is a process in which the ADP-ribose moiety of nicotinamide adenine dinucleotide is transferred to an acceptor amino acid. Five mammalian ADP-ribosyltransferases (ART1-ART5) have been cloned, and each ART is expressed in different tissues. ART5 (ADP-ribosyltransferase 5), also known as Ecto-ADP-ribosyltransferase 5, is a 292 amino acid secretory protein that is expressed in testis, heart, skeletal muscle and lymphoma. Functionally, ART5 is implicatetd to play a role in cell signaling and metabolism cascades. Two isoforms of ART5 exist as a result of alternative splicing events.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: ARTC5
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat