To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Arylsulfatase H
- Clonality:Polyclonal
- Gene ID:347527
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Antigen symbol:ARSH
- Conjugation:Alexa Fluor® 350
- ImmunoChemistry:Yes
- Size:100 µL
- Cross adsorption:No
- Form:liquid
- Antigen synonyms:Arylsulfatase H|ARSH_HUMAN.|ASH
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at –20 °C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Cat. no.:10667-104
- Supplier No.:BS-9101R-A350
Specifications
About this item
Sulfatases hydrolyze sulfate esters from sulfated steroids, carbohydrates, proteoglycans and glycolipids. They are involved in hormone biosynthesis, modulation of cell signaling and degradation of macromolecules. Arylsulfatase H, also known as ARSH, is a 562 amino acid protein that belongs to the sulfatase family of bone and cartilage matrix proteins. Localized to the plasma membrane, Arylsulfatase H uses calcium as a cofactor to hydrolyze sulfate esters. The gene encoding Arylsulfatase D maps to human chromosome X, which contains nearly 153 million base pairs and houses over 1,000 genes. In conjunction with chromosome Y, chromosome X is responsible for sex determination. There are a number of conditions related to an abnormal number and combination of sex chromosomes, some of which include Turner's syndrome, color blindness, hemophilia and Duchenne muscular dystrophy.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: ARSH
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human