To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Dipeptidyl-peptidase 6
- Clonality:Polyclonal
- Gene ID:1804
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Antigen symbol:DPP6
- Conjugation:FITC (Fluorescein Isothiocyanate)
- ImmunoChemistry:Yes
- Size:100 µL
- Cross adsorption:No
- Form:liquid
- Antigen synonyms:VF2|DPPX|MRD33
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at –20 °C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Cat. no.:10665-516
- Supplier No.:BS-9010R-FITC
Specifications
About this item
DPP6 is a Type-II serine proteinase of the clan SC. The clan SC proteinases have a catalytic triad of Ser-Asp-His, and like other Serine proteinases, the active site serine is in a Gly-Xaa-Ser-Xaa -Gly orientation. DPP6 has an Asp instead of Ser in the catalytic site. DPP6 is a member of a broader family of dipeptidyl peptidases including DPP4, FAP/Seprase, DPP2, DPP8, DPP9, DPP10, which have differing substrate specificity and tissue localizations. The surface-bound DPP6 is a homodimer, and cleavage of in the stalk region releases a shed form of DPP6. The shed is the form found in serum. DPP6 has been found in highest abundance in the brain, but also in the kidney, liver and lung.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: DPP6
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat