To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Breast Cancer Metastasis-suppressor 1
- Clonality:Polyclonal
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Antigen symbol:BRMS1
- Conjugation:Alexa Fluor® 555
- ImmunoChemistry:Yes
- Size:100 µL
- Cross adsorption:No
- Form:liquid
- Antigen synonyms:BRMS-1|AV003220|Breast cancer metastasis suppressor 1|MGC95128|AW554636.|Breast cancer metastasis-suppressor 1|BRMS1_HUMAN|DKFZp564A063
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at –20 °C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Cat. no.:10671-166
- Supplier No.:BS-8686R-A555
Specifications
About this item
Breast cancer metastasis-suppressor 1 (BRMS1) is 246 amino acid protein that acts as a mediator of metastasis suppression in several types of cancer including ovarian, lung, bladder, and murine mammary. BRMS1 mRNA is expressed in various tissues, including ovary, prostate, testis, and colon, but the protein is primarily detected in term placenta. BRMS1 suppresses metastasis without inhibiting tumorigenicity by modifying several metastasis-associated phenotypes. BRMS1 may participate in transcriptional regulation by binding to the mSin3/histone deacetylase complex. The expression of BRMS1 in certain cells increases connexin Cx43 expression and reduces connexin Cx32 expression. This produces a gap junction that increases intercellular communication, similar to those found in normal breast tissue. BRMS1 is stabilized by Hsp90 and may inhibit NF-â…¹B activity.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: BRMS1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat