Order Entry
ContactUsLinkComponent
Anti-FAM81A Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Anti-FAM81A Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Catalog #: 10671-070
Supplier:  Bioss
Anti-FAM81A Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Catalog #: 10671-070
Supplier:  Bioss
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Family With Sequence Similarity 81, Member A
  • Clonality:
    Polyclonal
  • Gene ID:
    145773
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Antigen symbol:
    FAM81A
  • Conjugation:
    Alexa Fluor® 350
  • ImmunoChemistry:
    Yes
  • Size:
    100 µL
  • Cross adsorption:
    No
  • Form:
    liquid
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Storage temperature:
    Store at –20 °C for 12 months
  • Concentration:
    1 μg/μl
  • Shipping temperature:
    4°C
  • Purification:
    Purified by Protein A
  • Cat. no.:
    10671-070
  • Supplier No.:
    BS-8654R-A350

Specifications

About this item

Encoding more than 700 genes, chromosome 15 is made up of approximately 106 million base pairs and is about 3% of the human genome. Angelman and Prader-Willi syndromes are associated with loss of function or deletion of genes in the 15q11-q13 region. In the case of Angelman syndrome, this loss is due to inactivity of the maternal 15q11-q13 encoded UBE3A gene in the brain by either chromosomal deletion or mutation. In cases of Prader-Willi syndrome, there is a partial or complete deletion of this region from the paternal copy of chromosome 15. Tay-Sachs disease is a lethal disorder associated with mutations of the HEXA gene, which is encoded by chromosome 15. Marfan syndrome is associated with chromosome 15 through the FBN1 gene. The FAM81A gene product has been provisionally designated FAM81A pending further characterization.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: FAM81A
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Customers Who Bought This Also Bought