To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:GRINL1A Complex Locus 1
- Clonality:Polyclonal
- Gene ID:145781
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Antigen symbol:GCOM1
- Conjugation:Cy7®
- ImmunoChemistry:Yes
- Size:100 µL
- Cross adsorption:No
- Form:liquid
- Antigen synonyms:MYZAP-POLR2M|Gcom2|GRINL1A|gcom|MYZAP
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at –20 °C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Cat. no.:10483-756
- Supplier No.:BS-8403R-CY7
Specifications
About this item
Glutamate receptors mediate most excitatory neurotransmission in the brain and play an important role in neural plasticity, neural development and neurodegeneration. Ionotropic glutamate receptors are categorized into NMDA receptors and kainate/AMPA receptors, both of which contain glutamate-gated, cation-specific ion channels. Synaptic and extrasynaptic NMDA receptors have been shown to have opposite effects on neuronal survival, CREB function and gene regulation. Gcom1 (GRINL1A complex locus protein 1), also known as GUP (GRINL1A upstream protein) and Gcom (GRINL1A combined protein), is a 466 amino acid protein that is a component of the GRINL1A complex transcription unit, which is thought to be involved in the modulation of glutamatergic neurotransmission through interaction with the NR1 subunit of the NMDA receptor. Gcom1 is expressed in small intestine, lung, liver, heart, skeletal muscle, testis and prostate and also colocalizes with NR1 in cortical and hippocampal neurons. There are eleven isoforms of Gcom1 that are produced as a result of alternative splicing events.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: GCOM1
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat