To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Ras-like without CAAX 1
- Clonality:Polyclonal
- Gene ID:6016
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Antigen symbol:RIT1
- Conjugation:Cy7®
- ImmunoChemistry:Yes
- Size:100 µL
- Cross adsorption:No
- Form:liquid
- Antigen synonyms:RIT|NS8|RIBB|ROC1
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at –20 °C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Cat. no.:10483-300
- Supplier No.:BS-8392R-CY7
Specifications
About this item
Plays a crucial role in coupling NGF stimulation to the activation of both EPHB2 and MAPK14 signaling pathways and in NGF-dependent neuronal differentiation.Neuronal activity dramatically increases the concentration of cytosolic Ca2+, which then serves as a second messenger to direct diverse cellular responses. Calmodulin is a primary mediator of Ca2+ signals in the nervous system. Ric, a protein related to the Ras subfamily of small GTPases, has the ability to bind calmodulin. In addition, two Ras-like human proteins, Rin and Rit (Ric-related gene expressed in many tissues), which are 71% and 66% identical to RIC respectively, share related G2 domains with Ric. While most members of the Ras subfamily are plasma membrane-associated and generally require a C-terminal isoprenyl group to bind to the plasma membrane, Rit and Rin lack the recognition signal for C-terminal prenylation. Transiently expressed Rit and Rin are plasma membrane-localized because both proteins contain a C-terminal cluster of basic amino acids, which provides a mechanism for membrane association. Rin binds calmodulin through a C-terminal binding motif. Rit and Ric are widely expressed, whereas expression of Rin is restricted to the neuron system. In conclusion, Rit and Rin define a novel subfamily of Ras-related proteins
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: RIT1
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat