To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Pantothenate Kinase 2
- Clonality:Polyclonal
- Gene ID:80025
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Antigen symbol:PANK2
- Conjugation:HRP (Horseradish Peroxidase)
- ImmunoChemistry:Yes
- Size:100 µL
- Western blot:Yes
- Cross adsorption:No
- Form:liquid
- Antigen synonyms:C20orf48|NBIA1|HARP|HSS|PKAN
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% Gentamicin. Store at 4°C for 12 months.
- Storage temperature:Store at –20 °C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Cat. no.:10482-240
- Supplier No.:BS-8338R-HRP
Specifications
About this item
Defects in PANK2 are the cause of neurodegeneration with brain iron accumulation type 1 (NBIA1); also known as pantothenate kinase-associated neurodegeneration (PKAN) or Hallervorden-Spatz syndrome (HSS). It is an autosomal recessive neurodegenerative disorder associated with iron accumulation in the brain, primarily in the basal ganglia. Clinical manifestations include progressive muscle spasticity, hyperreflexia, muscle rigidity, dystonia, dysarthria, and intellectual deterioration which progresses to severe dementia over several years. It is clinically classified into classic, atypical, and intermediate phenotypes. Classic forms present with onset in the first decade, rapid progression, loss of independent ambulation within 15 years. Atypical forms have onset in the second decade, slow progression, maintenance of independent ambulation up to 40 years later. Intermediate forms manifest onset in the first decade with slow progression or onset in the second decade with rapid progression. Patients with early onset tend to also develop pigmentary retinopathy, whereas those with later onset tend to also have speech disorders and psychiatric features. All patients have the 'eye of the tiger' sign on brain MRI.Defects in PANK2 are the cause of hypoprebetalipoproteinemia, acanthocytosis, retinitis pigmentosa, and pallidal degeneration (HARP). HARP is a rare syndrome with many clinical similarities to NBIA1.
Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500
Type: Primary
Antigen: PANK2
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat