To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Retinol Dehydrogenase 13
- Clonality:Polyclonal
- Gene ID:112724
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Antigen symbol:RDH13
- Conjugation:Cy5.5®
- ImmunoChemistry:Yes
- Size:100 µL
- Cross adsorption:No
- Form:liquid
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at –20 °C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Cat. no.:10482-186
- Supplier No.:BS-8333R-CY5.5
Specifications
About this item
RDH13, also known as all-trans and 9-cis retinol dehydrogenase 13 or SDR7C3, is a 331 amino acid mitochondrial protein belonging to the short-chain dehydrogenases/reductases (SDR) family. Widely expressed, mostly in eye, pancreas, placenta and lung, RDH13 localizes on the outer side of the inner mitochondrial membrane. Related to microsomal retinoid oxidoreductase RDH11, RDH13 is considered to be a major enzyme among the RDH family of proteins. Catalytically active, RDH13 recognizes retinoids as substrates and may function in retinoic acid production. RDH13 may function to protect the mitochondria against oxidative stress. Leber congenital amaurosis (LCA) type 3, an inherited autosomal recessive retinal disease, has been associated with defects of RDH13. LCA represents the most common genetic cause of congenital visual impairment in infants and children.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: RDH13
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat