To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Serpin Family B Member 13
- Clonality:Polyclonal
- Gene ID:5275
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Antigen symbol:SERPINB13
- Conjugation:Alexa Fluor® 488
- ImmunoChemistry:Yes
- Size:100 µL
- Cross adsorption:No
- Form:liquid
- Antigen synonyms:headpin|PI13|HUR7|HSHUR7SEQ
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at –20 °C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Cat. no.:10481-902
- Supplier No.:BS-8324R-A488
Specifications
About this item
Headpin is a skin-specific, UV-repressible serine proteinase inhibitor (serpin) belonging to the ovalbumin serpin family. Headpin is abundant in the human keratinocyte cell line HaCaT, and in lesional keratinocytes from psoriatic skin. Headpin downregulation occurs in squamous cell carcinoma of the oral cavity and in squamous cell carcinoma cell lines of the head and neck.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: SERPINB13
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat