To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:disrupted in renal carcinoma 2
- Clonality:Polyclonal
- Gene ID:84925
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Antigen symbol:DIRC2
- Conjugation:Cy3®
- ImmunoChemistry:Yes
- Size:100 µL
- Cross adsorption:No
- Form:liquid
- Antigen synonyms:RCC4
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at –20 °C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Cat. no.:10480-516
- Supplier No.:BS-8278R-CY3
Specifications
About this item
Belonging to the major facilitator superfamily, DIRC2 (disrupted in renal carcinoma protein 2) is a 478 amino acid multi-pass membrane protein that is primarily expressed in kidney proximal tubular cells. The genes encoding DIRC2 and DIRC3 are located at a translocation breakpoint which occurs frequently in individuals affected by familial renal cell carcinoma. Fusion transcripts that result from these translocations may affect normal protein function. This evidence suggests that, due to its chromosomal location, deregulation of the DIRC2 gene may cause haploinsufficiency and therefore result in the onset of tumor growth. There are two isoforms of DIRC2 which are produced as a result of alternative splicing events.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: DIRC2
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat