To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Collectin sub-family member 11
- Clonality:Polyclonal
- Gene ID:78989
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Antigen symbol:COLEC11
- Conjugation:FITC (Fluorescein Isothiocyanate)
- ImmunoChemistry:Yes
- Size:100 µL
- Cross adsorption:No
- Form:liquid
- Antigen synonyms:CL-K1-IIb|CL-K1-I|3MC2|CL-K1-IIa|CLK1|CL-K1-II
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at –20 °C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Cat. no.:10474-902
- Supplier No.:BS-8032R-FITC
Specifications
About this item
COLEC11 is a 271 amino acid C-type lectin protein that contains a collagen-like domain and a carbohydrate recognition domain, and plays an important role in host-defense. COLEC11 binds to various sugars and LPS (lipopolysaccharides), which include fucose but does not bind to glucose, hnRNP, Beta-1,3-Gal-T3 or mannose. COLEC11 is ubiquitously expressed in most tissues with high expression in kidney, liver, fetal liver, small intestine, thymus, spinal cord, placenta, adrenal gland, pancreas and several cell lines. COLEC11 is a secreted protein and all alternatively spliced isoforms of COLEC11 have oligomeric structures created through disulfide bonding.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: COLEC11
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype:
Reactivity: