To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Ankyrin Repeat Domain 11
- Clonality:Polyclonal
- Gene ID:29123
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Antigen symbol:ANKRD11
- Conjugation:Alexa Fluor® 647
- ImmunoChemistry:Yes
- Size:100 µL
- Cross adsorption:No
- Form:liquid
- Antigen synonyms:Ankyrin repeat domain containing protein 11|Ankyrin repeat containing cofactor 1|ANCO1|T13.|Ankyrin repeat domain 11|LZ16|ANCO 1
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at –20 °C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Cat. no.:10473-816
- Supplier No.:BS-7961R-A647
Specifications
About this item
Ankyrin is a membrane protein that mediates the attachment of the erythrocyte membrane skeleton to the plasma membrane and interacts with CD44 and inositol triphosphate. It contains three functional domains: a conserved N-terminal ankyrin repeat domain (ARD(consisting of 22–24 tandem repeats of 33 amino acids), a spectrin binding domain and a variably sized C-terminal regulatory domain. The ankyrin repeat is a 33-residue motif in proteins consisting of two alpha helices separated by loops. It has been studied using multiple sequence alignment to determine which conserved amino acid residues are critical for folding and stability. Ankyrin-repeat proteins have been associated with a number of human diseases; most notably, the cell cycle inhibitor p16 is associated with cancer and the Notch protein is a key component of cell signaling pathways whose intracellular repeat domain is disrupted in mutations that give rise to the neurological disorder known as CADASIL.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: ANKRD11
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat