To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Dual Specificity Phosphatase 13
- Clonality:Polyclonal
- Gene ID:51207
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Antigen symbol:DUSP13
- Conjugation:Cy5.5®
- ImmunoChemistry:Yes
- Size:100 µL
- Cross adsorption:No
- Form:liquid
- Antigen synonyms:MDSP|DUSP13A|BEDP|SKRP4|DUSP13B|TMDP
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at –20 °C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Cat. no.:10469-786
- Supplier No.:BS-7797R-CY5.5
Specifications
About this item
Members of the protein-tyrosine phosphatase superfamilycooperate with protein kinases to regulate cell proliferation anddifferentiation. This superfamily is separated into two familiesbased on the substrate that is dephosphorylated. One family, thedual specificity phosphatases (DSPs) acts on both phosphotyrosineand phosphoserine/threonine residues. This gene encodes differentbut related DSP proteins through the use of non-overlapping openreading frames, alternate splicing, and presumed differenttranscription promoters. Expression of the distinct proteins fromthis gene has been found to be tissue specific and the proteins maybe involved in postnatal development of specific tissues. A proteinencoded by the upstream ORF was found in skeletal muscle, whereasthe encoded protein from the downstream ORF was found only intestis. In mouse, a similar pattern of expression was found.Multiple alternatively spliced transcript variants were described,but the full-length sequence of only some were determined.[provided by RefSeq, Jul 2008].
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: DUSP13
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat