To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Septin 1
- Antigen symbol:SEPT1
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 488
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Size:100 µL
- Antigen synonyms:DIFF6|PNUTL3|LARP|SEPT1|SEP1|Septin-1
- Gene ID:1731
- Concentration:1 μg/μl
- Immunogen:160-210/367
- Purification:Purified by Protein A
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at –20 °C for 12 months
- Shipping temperature:4°C
- Form:liquid
- Cross adsorption:No
- Cat. no.:10469-390
- Supplier No.:BS-7769R-A488
Specifications
About this item
SEPT1 is a member of the septin family of GTPases. Members of this family are required for cytokinesis. SEPT1 is associated with the tau-based paired helical filament core, and may contribute to the formation of neurofibrillary tangles in Alzheimer's disease.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: SEPT1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat