To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Thyroid Hormone Receptor Interactor 1
- Clonality:Polyclonal
- Gene ID:5705
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Antigen symbol:TRIP1
- Conjugation:HRP (Horseradish Peroxidase)
- ImmunoChemistry:Yes
- Size:100 µL
- Western blot:Yes
- Cross adsorption:No
- Form:liquid
- Antigen synonyms:Thyroid Receptor Interactor 1|PSMC5|TRIP1|p45/S?g|macropain) 26S Subunit|TRIP-1|S?g1|Proteasome (prosome|Tat-binding protein homolog 10|S8|p45|TBP10|ATPase|5
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% Gentamicin. Store at 4°C for 12 months.
- Storage temperature:Store at –20 °C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Cat. no.:10468-232
- Supplier No.:BS-7731R-HRP
Specifications
About this item
Human SUG1 (also known as p45, Rpt6, Cim3 and PSMC5) is a component of the 19S regulatory subunit of the 26S proteosome complex. It is one of the AAA ATPas found in the 19S regulatory complex that is thought to participate in the unfolding of ubiquitinated proteins in an ATP dependent manner. It has also been shown to directly interact with the nucleotide excision repair protein XPB.
Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500
Type: Primary
Antigen: TRIP1
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat