To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Tumor Necrosis Factor Receptor Superfamily Member 11A
- Clonality:Polyclonal
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Antigen symbol:TNFRSF11A
- Conjugation:FITC (Fluorescein Isothiocyanate)
- ImmunoChemistry:Yes
- Size:100 µL
- Cross adsorption:No
- Form:liquid
- Antigen synonyms:ODFR|RANK|OFE|OSTS|FEO|LOH18CR1|OPTB7|CD265|PDB2|TRANCER
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at –20 °C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Cat. no.:10463-020
- Supplier No.:BS-7343R-FITC
Specifications
About this item
CD265 is a member of the tumor necrosis factor receptor (TNFR) family. Human and murine CD265 share 81% amino acid identity in their extracellular domains. CD265 is widely expressed, with highest levels in skeletal muscle, thymus, liver, colon, small intestine and adrenal gland. CD265 is also expressed in dendritic cells. RANK and RANK ligand (RANKL) are important regulators of interactions between T cells and dendritic cells. RANK is the essential signaling receptor for osteoclast differentiation factor in osteoclastogenesis. Multiple tumor necrosis factor receptor-associated factors (TRAFs) are involved in the signaling of CD265. TRANCE (TNF-related activation-induced cytokines, also known as RANK ligand, osteoprotegerin ligand and osteoclast differentiation factor) is the ligand for CD265. The biological functions mediated by RANK include activation of NFkappaB and cjun N-terminal kinase, enhancement of T cell growth and dendritic cell function, induction of osteoclastogenesis and lymph node organogenesis. The soluble form of CD265 is able to block TRANCE induced biological activity. The binding of anti-CD265 to cell surface CD265 triggers signal transduction and induces CD265 mediated bioactivity.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: TNFRSF11A
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat