To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Oxysterol Binding Protein 2
- Clonality:Polyclonal
- Gene ID:23762
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Antigen symbol:OSBP2
- Conjugation:Cy7®
- ImmunoChemistry:Yes
- Size:100 µL
- Cross adsorption:No
- Form:liquid
- Antigen synonyms:ORP-4|ORP4|OSBPL4|HLM|OSBPL1
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Cat. no.:10449-836
- Supplier No.:BS-6560R-CY7
Specifications
About this item
The Oxysterol-binding protein (OSBP) family of proteins consist of OSBP (OSBP1) and OSBP2 (ORP-4), which share a high overall similarity. OSBPs are involved in lipid metabolism and signal transduction, as well as vesicle transport, and can translocate to the periphery of Golgi membranes when they are bound to oxysterols. The OSBP protein transports sterols from lysosomes to the nucleus, where sterols downregulate the genes for HMG synthetase, HMG-CoA reductase and the low density lipoprotein receptor (LDLR). OSBP localizes to the cytosol and is widely expressed, while OSBP2 is mainly detected in testis, retina and fetal liver. The extracellular signal-regulated kinase (ERK) signaling pathway is controlled by OSBP via its cholesterol-binding properties. OSBP binds with a high affinity to 25-hydroxy-cholesterol (25-HC), a suppressor of cholesterol synthesis gene transcription in cultured cells.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: OSBP2
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat