To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Arachidonate 15-Lipoxygenase Type B
- Clonality:Polyclonal
- Gene ID:247
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Antigen symbol:ALOX15B
- Conjugation:Cy7®
- ImmunoChemistry:Yes
- Size:100 µL
- Cross adsorption:No
- Form:liquid
- Antigen synonyms:15-LOX-2
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Immunogen:5-55/676
- Purification:Purified by Protein A
- Cat. no.:10435-544
- Supplier No.:BS-6335R-CY7
Specifications
About this item
Non-heme iron-containing dioxygenase that catalyzes the stereo-specific peroxidation of free and esterified polyunsaturated fatty acids generating a spectrum of bioactive lipid mediators. Converts arachidonic acid to 15S-hydroperoxyeicosatetraenoic acid/(15S)-HPETE. Also acts on linoleic acid to produce 13-hydroxyoctadecadienoic acid/13-HPODE. Has no detectable 8S-lipoxygenase activity but reacts with (8S)-HPETE to produce (8S,15S)-diHPETE. May regulate progression through the cell cycle and cell proliferation. May also regulate cytokine secretion by macrophages and therefore play a role in the immune response. May also regulate macrophage differentiation into proatherogenic foam cells.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: ALOX15B
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human