To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:ADAM Metallopeptidase Domain 11
- Clonality:Polyclonal
- Gene ID:4185
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Antigen symbol:ADAM11
- Conjugation:FITC (Fluorescein Isothiocyanate)
- ImmunoChemistry:Yes
- Size:100 µL
- Cross adsorption:No
- Form:liquid
- Antigen synonyms:Metalloproteinase like|disintegrin like|MDC|A disintegrin and metalloproteinase domain 11|ADA11_HUMAN|and cysteine rich protein.
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Cat. no.:10424-958
- Supplier No.:BS-5846R-FITC
Specifications
About this item
ADAM11 was first described as MDC (Metalloproteinase-like disintergin-like cysteine-rich protein) from analysis of human brain libraries, in search of brain-specific proteins. Two splice variants with different carboxyterminal ends were described. The message was found only in the brain in this publication. Another group identified ADAM11 in the human brain, where ADAM11 was thought to be involved in cell migration and spatial patterning. ADAM11 was mapped to 17q21.3, a region of interest for breast cancer, and mutations in ADAM11 are associated with some breast cancers. Retinoic acid caused a doubling in ADAM11 message levels over 24 hours in NT2/D1 cells, a human embryonic carcinoma cell line. ADAM11 null mutant mice have deficits in spatial learning and motor coordination, although they did have normal cell migration and differentiation during development. ADAM11 is a member of the ADAMs family (A Disintegrin And Metalloproteinase), but does not contain the canonical HExxHxxxxH zinc-binding metalloproteinase catalytic site. The domain structure of the full-length ADAM11 includes a signal sequence, propeptide domain, metalloproteinase-like domain, disintegrin-like domain, cys-rich domain, EGF-like domain, a spacer region, then the transmembrane domain and a short cytoplasmic domain.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: ADAM11
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat