To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Integrin Alpha 5
- Clonality:Polyclonal
- Gene ID:3685
3694 - Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Antigen symbol:ITGA5
- Conjugation:Alexa Fluor® 647
- ImmunoChemistry:Yes
- Size:100 µL
- Cross adsorption:No
- Form:liquid
- Antigen synonyms:MSK8|Vitronectin Receptor Subunit Alpha|CD51|ITGAV|Vitronectin Receptor|VNRA|VTNR
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Cat. no.:10423-322
- Supplier No.:BS-5791R-A647
Specifications
About this item
Integrins are heterodimeric cell surface receptors composed of alpha and beta subunits, which mediate cell-cell and cell-extracellular matrix attachments. Aberrant integrin expression has been found in many epithelial tumours. Changes in integrin expression have been shown to be important for the growth and early metastatic capacity of melanoma cells. Integrin alpha-v beta-6 is upregulated in cancers and during tissue remodelling but is absent from resting adult tissues. Integrin alpha-v beta-6 promotes invasion and correlates with poor survival and therefore makes a promising therapeutic target.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: ITGAV
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host:
Isotype:
Reactivity: