To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:BRICK1, SCAR/WAVE actin-nucleating complex subunit
- Clonality:Polyclonal
- Gene ID:55845
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Antigen symbol:BRK1
- Conjugation:Alexa Fluor® 647
- ImmunoChemistry:Yes
- Size:100 µL
- Cross adsorption:No
- Form:liquid
- Antigen synonyms:HSPC300|C3orf10|MDS027|hHBrk1
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Cat. no.:10409-048
- Supplier No.:BS-5124R-A647
Specifications
About this item
HSPC300 (hematopoietic stem cell protein 300) is also known as probable protein BRICK1 or C3orf10 (chromosome 3 open reading frame 10) and is a 75 amino acid protein that is expressed as two isoforms and localizes to both the cytoplasm and the cytoskeleton. HSPC300 is thought to regulate cytoskeletal organization and Actin polymerization. Free HSPC300 exists as homotrimers prior to its incorporation into the WAVE complex. The WAVE complex includes five proteins, one of which is HSPC300, that regulate the ARC (Arp2/3 complex) which is responsible for Actin nucleation and is Rac 1-dependent. Because HSPC300 is a highly conserved subunit of the WAVE complex across many species, it is thought to have the same or similar functions in many different organisms. In Drosophila, the WAVE/ARC pathway may affect the development of the nervous system. HSPC300 is thought to localize to axons of the central nervous system of Drosophila embryos and thus may also be involved in axonogenesis. In addition, HSPC300 is thought to be necessary for synaptic morphogenesis by motoneurons. In mice, the knockout of the WAVE complex leads to learning and memory defects, and it is therefore hypothesized that HSPC300 may also be involved in cognitive functions. Genetic depletion of HSPC300 results in cytoskeletal abnormalities and prevents cytokinesis of cells, suggesting that decreased levels of HSPC300 may be associated with tumor suppression.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: BRK1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat