To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Poly(A) Polymerase Gamma
- Clonality:Polyclonal
- Gene ID:64895
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Antigen symbol:PAPOG
- Conjugation:Cy7®
- ImmunoChemistry:Yes
- Size:100 µL
- Cross adsorption:No
- Form:liquid
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Immunogen:105-155/736
- Purification:Purified by Protein A
- Cat. no.:10399-780
- Supplier No.:BS-4968R-CY7
Specifications
About this item
Responsible for the post-transcriptional adenylation of the 3'-terminal of mRNA precursors and several small RNAs including signal recognition particle (SRP) RNA, nuclear 7SK RNA, U2 small nuclear RNA, and ribosomal 5S RNA.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: PAPOG
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat