To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Mal, T-Cell Differentiation Protein
- Clonality:Polyclonal
- Gene ID:4118
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat
- Antigen symbol:MAL
- Conjugation:FITC (Fluorescein Isothiocyanate)
- ImmunoChemistry:Yes
- Size:100 µL
- Cross adsorption:No
- Form:liquid
- Antigen synonyms:T-Lymphocyte Maturation-Associated Protein
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Immunogen:115-125/153
- Purification:Purified by Protein A
- Cat. no.:10398-988
- Supplier No.:BS-4835R-FITC
Specifications
About this item
Could be an important component in vesicular trafficking cycling between the Golgi complex and the apical plasma membrane. Could be involved in myelin biogenesis and/or myelin function.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: MAL
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat