To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Neural Precursor Cell Expressed, Developmentally Down-Regulated 8
- Clonality:Polyclonal
- Gene ID:4738
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Antigen symbol:NEDD8
- Conjugation:Alexa Fluor® 350
- ImmunoChemistry:Yes
- Size:100 µL
- Cross adsorption:No
- Form:liquid
- Antigen synonyms:NEDD 8|NED8|Neural precursor cell expressed developmentally down regulated gene 8|Neddylin|Ubiquitin like protein Nedd 8|Neural precursor cell expressed developmentally down regulated 8|Ubiquitin like protein Nedd8.|Rub1
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Cat. no.:10388-146
- Supplier No.:BS-3812R-A350
Specifications
About this item
Ubiquitin like proteins fall into two classes: the first class, ubiquitin-like modifiers (UBLs) function as modifiers in a manner analogous to that of ubiquitin. Examples of UBLs are SUMO, Rub1 (also called Nedd8), Apg12 and Hub1. Proteins of the second class include Parkin, RAD23 and DSK2, are designated ubiquitin domain proteins (UDPs). These proteins contain domains that are related to ubiquitin but are otherwise unrelated to each other. In contrast to UBLs, UDPs are not proteolytically processed or conjugated to other proteins. Rub1 and the corresponding human homolog NEDD8 are activated by the E1 ubiquitin activating enzyme UBA2, that forms isopeptide linkages between thio esters. NEDD8 shows 80% homology to ubiquitin. The best known targets of Rub1 modification are members of the cullin family. Cullins are subunits of an E3 ubiquitin ligase complex called the Skp1 /Cul1/Cdc53 F box (SCF). The SCF promotes transfer of ubiquitin from a ubiquitin conjugating enzyme (E2) to the target protein. Rub1 modification may regulate SCF function or localization.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: NEDD8
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat