To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:ST6 Beta-Galactosamide Alpha-2,6-Sialyltranferase 1
- Clonality:Polyclonal
- Gene ID:6480
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Antigen symbol:ST6GAL1
- Conjugation:Alexa Fluor® 488
- ImmunoChemistry:Yes
- Size:100 µL
- Cross adsorption:No
- Form:liquid
- Antigen synonyms:B-cell antigen CD75|6-sialyltransferase 1|alpha 2|ST6GalI|6-sialyltransferase|ST6Gal I|CMP-N-acetylneuraminate beta-galactosamide alpha-2|6-ST 1|ST6N|beta-galactoside alpha-2|sialyltransferase 1|CMP-N-acetylneuraminate-beta-galactosamide-alpha-2|Sialyltransferase 1|SIAT1
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Immunogen:325-375/406
- Purification:Purified by Protein A
- Cat. no.:10394-866
- Supplier No.:BS-3793R-A488
Specifications
About this item
The protein encoded by this gene is a type II membrane protein that catalyzes the transfer of sialic acid from CMP sialic acid to galactose-containing substrates. The encoded protein, which is normally found in the Golgi apparatus but which can be proteolytically processed to a soluble form, is involved in the generation of the cell surface carbohydrate determinants and differentiation antigens HB6, CDw75, and CD76. This protein is a member of glycosyltransferase family 29. Three transcript variants encoding two different isoforms have been found for this gene.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: ST6GAL1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat