To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 13
- Clonality:Polyclonal
- Gene ID:51079
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Antigen symbol:NDUFA13
- Conjugation:HRP (Horseradish Peroxidase)
- ImmunoChemistry:Yes
- Size:100 µL
- Western blot:Yes
- Cross adsorption:No
- Form:liquid
- Antigen synonyms:CGI-39|GRIM19|CDA016|GRIM-19|B16.6
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% Gentamicin. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Cat. no.:10375-568
- Supplier No.:BS-3594R-HRP
Specifications
About this item
A novel gene, Genes associated with Retinoid IFN induced Mortality (GRIM) GRIM19 gene was identified. Antisense expression of GRIM19 confers a strong resistance against IFN/RA induced death by reducing the intracellular levels of GRIM19 protein. Overexpression of GRIM19 enhances cell death in response to IFN/RA. GRIM19 is primarily a nuclear protein whose expression is induced by the IFN/RA combination. These data indicate that GRIM19 is a novel cell death regulatory molecule.
Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500
Type: Primary
Antigen: GRIM19
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat