To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Kinesin Family member 17
- Clonality:Polyclonal
- Gene ID:57576
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Antigen symbol:KIF17
- Conjugation:Alexa Fluor® 647
- ImmunoChemistry:Yes
- Size:100 µL
- Cross adsorption:No
- Form:liquid
- Antigen synonyms:KIF17B|KLP-2|KIF3X|OSM-3
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Cat. no.:10374-784
- Supplier No.:BS-3527R-A647
Specifications
About this item
The kinesin superfamily of proteins (KIFs) consists of a class of microtubule-dependent motors that play a major role in many cellular and developmental functions, including organelle transport, mitosis, meiosis, and possibly long-range signaling in neurons. The kinesin proteins are involved in organelle transport and are primarily associated with anterograde transport of vesicles and organelles in neurons, epithelial cells, and melanosomes with bidirectional transport of mitochondria. They also mediate transport between the endoplasmic reticulum (ER) and the Golgi complex. In neurons, kinesin motors conduct vesicular transport, such as of synaptic vesicle components to axons and of neurotransmitter receptors to dendrites. KIF17 belongs to the functionally diverse subgroup of the kinesin superfamily characterized by a N-terminal motor domain (N-IV class), that includes the KIF3 motor protein. KIF17 is specifically expressed in the brain, present in abundance in the gray matter, particularly in the hippocampus and cerebral cortex, but not in the white matter such as the optic nerve.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: KIF17
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat