To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Fibrinogen Beta Chain
- Clonality:Polyclonal
- Gene ID:2244
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse
- Antigen symbol:FGB
- Conjugation:Cy3®
- ImmunoChemistry:Yes
- Size:100 µL
- Cross adsorption:No
- Form:liquid
- Antigen synonyms:MGC104327|FGB ,fibrinogen beta chain|MGC120405
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Cat. no.:10374-642
- Supplier No.:BS-3514R-CY3
Specifications
About this item
D-DIMER is A fibrin degradation fragment or product that is produced by the action of plasmin on fibrin in the clot dissolution process.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: D-DIMER
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse