To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Discs, Large Homolog 4 (Drosophila)
- Clonality:Polyclonal
- Gene ID:1742
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Antigen symbol:DLG4
- Conjugation:Alexa Fluor® 555
- ImmunoChemistry:Yes
- Size:100 µL
- Cross adsorption:No
- Form:liquid
- Antigen synonyms:Tax interaction protein 15|SAP-90|discs large homolog 4|SAP90|large homolog 4 (Drosophila)|FLJ98574|DLG4|synapse-associated protein 90|discs|PSD95|post-synaptic density protein 95|FLJ97752
- Modification:phospho-Tyr236/Tyr240
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Cat. no.:10367-794
- Supplier No.:BS-3349R-A555
Specifications
About this item
postsynaptic density protein 95 is a molecular scaffolding protein that binds and clusters N-methyl-D-aspartate receptors at neuronal synapses; may be involved in guanine nucleotide-mediated signal transduction pathway. This gene encodes a member of the membrane-associated guanylate kinase (MAGUK) family. It heteromultimerizes with DLG2. With DLG2 it is recruited into the same NMDA receptor and potassium channel clusters. These two MAGUK proteins may interact at postsynaptic sites to form a multimeric scaffold for the clustering of receptors, ion channels, and associated signaling proteins.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: DLG4
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat