To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Interleukin 12 Receptor Beta 1
- Clonality:Polyclonal
- Gene ID:16161
- Host:Rabbit
- Isotype:IgG
- Reactivity:Rat,Mouse
- Antigen symbol:IL12RB1
- Conjugation:Cy7®
- Flow cytometry:Yes
- ImmunoChemistry:Yes
- Size:100 µL
- Cross adsorption:No
- Form:liquid
- Antigen synonyms:CD212|IL12RB|beta 1|IL-12R-BETA1|IL-12 receptor beta component|MGC34454|IL12RB1|interleukin-12 receptor beta-1 chain|interleukin 12 receptor
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Immunogen:420-470/668
- Purification:Purified by Protein A
- Cat. no.:10342-558
- Supplier No.:BS-2603R-CY7
Specifications
About this item
Functions as an interleukin receptor which binds interleukin-12 with low affinity and is involved in IL12 transduction. Associated with IL12RB2 it forms a functional, high affinity receptor for IL12. Associates also with IL23R to form the interleukin-23 receptor which functions in IL23 signal transduction probably through activation of the Jak-Stat signaling cascade.
Recommended Dilutions: Flow Cytometry: 1:20-100; IF(IHC-P): 1:50-200; IF(IHC-F)(1:50-200
Type: Primary
Antigen: IL12RB1
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat