To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Kruppel-like Factor 5
- Clonality:Polyclonal
- Gene ID:688
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Antigen symbol:KLF5
- Conjugation:Alexa Fluor® 350
- ImmunoChemistry:Yes
- Size:100 µL
- Cross adsorption:No
- Form:liquid
- Antigen synonyms:IKLF|CKLF|BTEB2
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Immunogen:80-130/457
- Purification:Purified by Protein A
- Cat. no.:10334-286
- Supplier No.:BS-2385R-A350
Specifications
About this item
The kinesin motor proteins include at least two forms of conventional kinesin encoded by different genes and designated as ubiquitous kinesin, which is expressed in all cells and tissues, or neuronal kinesin, which is expressed exclusively in neural cells. Kinesin is a microtubule associated protein comprised of three different structural domains. A considerable globular N-terminal domain regulates the hydrolysis of ATP and also microtubule binding while central coiled-coil domains promote heavy chain dimerization. Lastly, small globular C-terminal domains interact with kinesin light chains, membranous organelles and vesicles. Expression of ubiquitous kinesin heavy chain, also designated UKHC, is found subcellularly in areas of heavy vesicular trafficking such as the microtubule pathways of neural cells and also the Golgi of non-neural cell types.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: KLF5
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat