To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Lysosomal-associated membrane protein 2
- Antigen symbol:LAMP2
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 647
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Size:100 µL
- Antigen synonyms:LAMP-2|LGP-B|Lysosome-associated membrane protein 2|CD107 antigen-like family member B|LAMP2|Lysosome-associated membrane glycoprotein 2|CD107b|Lysosomal membrane glycoprotein type B
- Gene ID:3920
- Concentration:1 μg/μl
- Immunogen:305-355/410
- Purification:Purified by Protein A
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Shipping temperature:4°C
- Form:liquid
- Cross adsorption:No
- Cat. no.:10333-972
- Supplier No.:BS-2379R-A647
Specifications
About this item
Implicated in tumor cell metastasis. May function in protection of the lysosomal membrane from autodigestion, maintenance of the acidic environment of the lysosome, adhesion when expressed on the cell surface (plasma membrane), and inter- and intracellular signal transduction. Protects cells from the toxic effects of methylating mutagens.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: LAMP2
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat