To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:small Cajal body-specific RNA 22
- Clonality:Polyclonal
- Gene ID:677770
- Host:Rabbit
- Isotype:IgG
- Reactivity:Plant
- Antigen symbol:SCARNA22
- Conjugation:FITC (Fluorescein Isothiocyanate)
- ImmunoChemistry:Yes
- Size:100 µL
- Cross adsorption:No
- Form:liquid
- Antigen synonyms:ACA11
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Cat. no.:10328-556
- Supplier No.:BS-2249R-FITC
Specifications
About this item
In plant cells, the vacuole functions as a major calcium store. Although a calmodulin-regulated Ca2+-ATPase (ACA4) is known to be present in prevacuolar compartments, the presence of an ACA-type Ca2+-ATPase in the mature vacuole of a plant cell has not been verified. Here we provide evidence that ACA11 localizes to the vacuole membrane. ACA11 tagged with GFP was expressed in stable transgenic plants, and visualized in root cells and protoplasts by confocal microscopy. A Ca2+-ATPase function for ACA11 was confirmed by complementation of yeast mutants.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: SCARNA22
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Plant