Specifications
- Description:Determination of the Nitrite and Nitrate Concentrations in Water, 40 Tests
- Cat. no.:470219-152
- Includes:Instructions
Specifications
About this item
With this kit, students test water samples the same way analysts in the fields do.
- Test identifies nitrite and nitrate in fresh water samples
- Simple colorimetric test
- Clear results
- Required materials included
- Aligned with GLOBE protocols
This kit is designed for an analyst to perform a simple colorimetric test to determine the nitrite and nitrate concentrations of fresh water samples. The reaction yields an easily identifiable color transition that students can compare with the colors on the color chart to determine the concentrations.
Identified nitrite and nitrate values are 0.0, 0.25, 0.5, 1.0, 2.0, 4.0, 6.0, 8.0, and 10.0 ppm. This kit contains everything an analyst would need to perform a water test in a laboratory setting or out in the field.