About this item
Offering exceptional cut protection in a form-fitting ultralight weight glove with enhanced grip performance, the HyFlex® 11-541 epitomizes the convergence of protection and performance.
- Enhanced abrasion resistance
- Excellent grip
- Superior tactile sensitivity
- Unparalleled dexterity
It features an 18G ultra-lightweight liner with INTERCEPT TECHNOLOGY® yarns that provides EN Cut Level 4 (ANSI Cut Level 4) protection.
This glove's ultralight weight enables unparalleled dexterity and refined mobility for bare hand-like tactility. ZONZ™ comfort fit technology uses selected yarns and varying stitch designs to tailor overall glove fit to enhance hand movement for higher dexterity and reduced fatigue. Its silicone-free design is paint- and finish-process-friendly and dirt-masking palm allows longer wear times.
INTERCEPT TECHNOLOGY® yarn has been designed to perform at or exceed industry standards. This Ansell proprietary technology offers best-in-class cut protection and increased comfort with dexterity. Palm coated protection provides grip while not limiting comfort, and back-of-hand breathability. ANSELL GRIP™ technology is a coating treatment that minimizes the force required to grip oily or wet tools or materials, reducing hand and arm fatigue and improving control, safety and productivity. The HyFlex® 11-541 glove also features FORTIX™ technology which applies a thin, resilient and breathable nitrile foam coating, which extends gloves working life and improves comfort in abrasive applications.
For use in the aerospace, automotive, machinery and equipment, and metal fabrication industries for assembly of small sharp parts, handling small sharp metal parts, light duty stamping and maintenance.
Specifications
- Catalog No:
- CA10002-004
- CA10002-074
- CA10002-076
- CA10002-078
- CA10002-080
- CA10002-082
- Ambidextrous/Hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Latex/Latex-free:
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Material:
- Nitrile
- Nitrile
- Nitrile
- Nitrile
- Nitrile
- Nitrile
- Cuff style:
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- EN norm:
- —
- —
- —
- —
- —
- —
- Anti-static:
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes