ADC® Pocket Ophthalmoscope Set
Catalog #: 470119-692
Supplier: AMERICAN DIAGNOSTIC CORP CP
Specifications
- Description:Pocket Ophthalmoscope Set
- Cat. no.:470119-692
- Supplier No.:W51514
Specifications
About this item
This pocket ophthalmoscope set features a 2.5 V halogen light source interchangeable “AA” metal battery handle.
- Impact resistant head
Pocket clip with integral on/off slide switch for ambidextrous use. Lifetime quality performance guarantee on optics.
Customers Who Bought This Also Bought