(2S,3aS,7aS)-2-Carboxyoctahydroindole
Supplier: Bachem Americas
Certificates
About this item
Synonyms: (2S,3aS,7aS)-Octahydro-1H-indole-2-carboxylic acid
Formula:C₉H₁₅NO₂
MW:169.22 g/mol
MDL Number:MFCD07782125
CAS Number:80875-98-5
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
:M.W. 169.22 g/mol
:(2S,3aS,7aS)-2-Carboxyoctahydroindole
Storage Temperature:Freezer
Customers Who Bought This Also Bought