About this item
The Motic BA310 is designed for the rigors of daily routine work
The Motic BA310 features a Siedentopf style head inclined at 30° and featuring full 360° rotation for comfortable viewing over extended periods. Also included are Widefield high eyepoint eyepieces, CCIS EF-N Plan Achromat objectives (4x, 10x, 40x S, and 100x S-oil), coaxial coarse and fine focusing adjustment, a built-in low position coaxial mechanical stage (right-hand control), and a focusable Abbe condenser with iris diaphragm and slot. All inclusions help ensure high quality, reproducible images. Additional optional contrast methods such as phase contrast, polarization, and darkfield (as well as discussion/teaching devices) ensure that this model will offer long term functionality to all user levels.
The requirements of any routine microscope demand careful attention to detail on every microscope detail. The BA310 provides just this, with a powerful adjustable light source to meet both delicate and detailed lighting requirements for all sample types, and full Koehler illumination to help in highlighting even the weakest stained specimens. The CCIS EF-N Plan Achromat objectives provide optimal image contrast through engineered multi-layer lens coatings to give superb image clarity and flatness of field.
Specifications
- Catalog No:
- CA89168-474
- CA89168-452
- CA89168-470
- CA89168-450
- Eyepieces:
- Objectives:
- Magnification:
- Condenser:
- Illumination:
- Electrical:
- Optical design:
- Accessories included: