PVC Dropper Tip, Wheaton
Catalog #: 16300-061
Supplier: DWK Life Sciences (KIMBLE)
Small Business Enterprise
Specifications
- Color:Clear
- Material:PVC
- Size:23 Dia. x 44 H mm
- Supplier No.:224080
Specifications
About this item
This PVC Dropper Tip attaches easily to serum bottles with a 20mm O.D
finish. The dropper tip is made from PVC, with a PE cap. It dispenses a 40μL drop using distilled water. This is an easy and effective method of converting a serum bottle to a dropping bottle.
Customers Who Bought This Also Bought