To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:fibroblast growth factor 13
- Clonality:Monoclonal
- Clone:S235-22
- Gene ID:2258
- Host:Mouse
- Reactivity:Human,Rat,Mouse
- Antigen symbol:FGF13
- Conjugation:FITC (Fluorescein Isothiocyanate)
- ELISA:Yes
- ImmunoChemistry:Yes
- Size:100 µg
- Western blot:Yes
- Epitope:N-Terminal
- Format:IgG2b
- Form:Liquid (sterile filtered)
- Antigen synonyms:FGF2|FHF-2|FHF2|FGF-13
- Concentration:1mg/mL by UV absorbance at 280 nm
- Shipping temperature:Wet Ice
- Immunogen:Anti-FGFA/FHFA (pan) Antibody was produced by repeated immunization of mice with a synthetic peptide containing amino acids 2-18 of human FHF2A.
- Purification:Anti-FGFA/FHFA (pan) Antibody was purified from concentrated tissue culture supernate by Protein G chromatography. BLAST analysis suggests that it is 94% identical to mouse, 100% identical to rat and >80% identical to FGF12A/FHF1A, FGF14A/FHF4A, and FGF11A/FHF3A.
- Cat. no.:10801-354
- Supplier No.:200-302-W62
Specifications
About this item
FGF13(Fibroblast growth factor 13), also called FHF2 is a protein that in humans is encoded by the FGF13 gene.The protein encoded by this gene is a member of the fibroblast growth factor (FGF) family. FGF13is a large gene, extending over approximately 200 kb in Xq26.3, and contains at least 7 exons. By cytogenetic, FISH, and database analysis, Gecz et al. (1999) localized the FGF13 gene within a 400-kb duplication interval on chromosome Xq26.3. FGF family members possess broad mitogenic and cell survival activities, and are involved in a variety of biological processes, including embryonic development, cell growth, morphogenesis, tissue repair, tumor growth, and invasion. Other areas of interests and use for researchers include cancer, borjeson-forssman-lehmann syndrome, regulation of actin cytoskeleton pathways, apoptosis, GPCR pathways, TGF-Beta pathways, and Rho familty GTPase pathways. This gene is located to a region associated with Borjeson-Forssman-Lehmann syndrome (BFLS), a syndromal X-linked mental retardation, which suggests it may be a candidate gene for familial cases of the BFL syndrome. The function of this gene has not yet been determined. Two alternatively spliced transcripts encoding different isoforms have been described for this gene.
Recommended Dilutions: ELISA: 1:10,000; Immunohistochemsitry: User Optimized; Western Blot: 1:1000; 0.1% (w/v) Sodium Azide
Type: Primary
Antigen: FHF2A
Clonality: Monoclonal
Clone: S235-22
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope: N-terminal
Host: Mouse
Isotype:
Reactivity: Human, Rat, Mouse