Skinfold Caliper, 3B Scientific
Catalog #: 470119-588
Supplier: American 3B Scientific
Specifications
- Description:Skinfold Caliper
- Cat. no.:470119-588
- Supplier No.:W50171
Specifications
About this item
This lightweight aluminum skinfold caliper includes a measurement arm with "floating tips" that under constant pressure assures accurate and reproducible results.
Easy to read 60 mm scale.
Includes booklet, tables and caliper carrying case.
Customers Who Bought This Also Bought