Algae and Environmental Problems Slide Set
Catalog #: 470177-044
Supplier: Avantor
Specifications
- Description:Algae and Environmental Problems Slide Set with 25 Slides
- Cat. no.:470177-044
- Supplier No.:WARD470177-044
Specifications
About this item
Identify indicator protists associated with polluted conditions
- Identifying features clearly distinguishable
- Rigorous quality control standards
Ordering information: Set Includes: Batrachosperum, Mixed Dinoflagellates and Diatoms, Synura, Navicula, Volvox, Ulothrix, Stigeoclonium, Spirogyra, Scenedesmus, Pediastrum, Pandorina, Oedogonium, Stuarstrum, Closterium, Chlorella, Chlamydomonas, Ankistrodesmus, Rivularia, Oscillatoria, Anacystis, Anabaena.
Customers Who Bought This Also Bought