Corning® CellBIND® 384-Well, Clear Treated Microplate, Corning
Catalog #: 10185-092
Supplier: Corning
Specifications
- Sterility:Sterile
- Color:Clear
- Description:CellBIND® Treated Microplate
- Environmentally Preferable:Low Manufacturing Impact
- No. of wells:384-Well
- Environmental certification:RECs, ISO 14001 - Kennebunk
- Cat. no.:10185-092
- Surface Treatment:CellBIND®
- Supplier No.:4579
Specifications
About this item
The Corning® CellBIND® surface enhances cell attachment under difficult conditions, such as reduced-serum or serum-free medium
This enhancement results in higher cell yields.
Customers Who Bought This Also Bought